3'-Aminoacetophenone oxime 95%

Supplier: Ambeed
Danger

A984423-1G A984423-250MG
77106-502EA 94.43 USD
77106-502 77106-504
3'-Aminoacetophenone oxime 95%
3'-Aminoacetophenone oxime

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.17 g/mol
MDL Number: MFCD18827945
CAS Number: 6011-18-3
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR