2,5-Dibromobenzonitrile 95%

Supplier: Ambeed
Warning

A978549-1G A978549-250MG A978549-5G
77106-490EA 31 USD
77106-490 77106-492 77106-494
2,5-Dibromobenzonitrile 95%
2,5-Dibromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Br₂N
MDL Number: MFCD00672994
CAS Number: 57381-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR