4-(3-Ethylthioureido)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)thiourea

A967511-1G A967511-250MG A967511-5G
77106-476EA 108 USD
77106-476 77106-478 77106-480
4-(3-Ethylthioureido)phenylboronic acid pinacol ester 97%
4-(3-Ethylthioureido)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BN₂O₂S
MDL Number: MFCD09972181
CAS Number: 1073353-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR