2,4-Dichlorobenzene-1,3-diol 95%

Supplier: Ambeed
Warning

A966409-10G A966409-250MG A966409-1G
77106-466EA 228 USD
77106-466 77106-468 77686-993
2,4-Dichlorobenzene-1,3-diol 95%
2,4-Dichlorobenzene-1,3-diol

Precaución: for Research Use Only

Formula: C₆H₄Cl₂O₂
MW: 179.001 g/mol
MDL Number: MFCD13152136
CAS Number: 16606-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR