L-Kynurenine 98%

Supplier: Ambeed
Warning

Synonyms: β-Anthraniloyl-L-alanine, (S)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, Kynurenine

A965360-100MG A965360-1G A965360-250MG
77106-460EA 75.33 USD
77106-460 77106-462 77106-464
L-Kynurenine 98%
L-Kynurenine

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
MDL Number: MFCD00069912
CAS Number: 2922-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR