3,3'-Bis(trifluoromethyl)benzophenone 98%

Supplier: Ambeed
Warning

Synonyms: Bis(3-(trifluoromethyl)phenyl)methanone

A963581-1G A963581-250MG A963581-5G
77106-454EA 21 USD
77106-454 77106-456 77106-458
3,3'-Bis(trifluoromethyl)benzophenone 98%
3,3'-Bis(trifluoromethyl)benzophenone

Precaución: for Research Use Only

Formula: C₁₅H₈F₆O
MDL Number: MFCD00000390
CAS Number: 1868-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR