2-Chloro-3-methyl-4-(methylsulfonyl)benzoic acid 95%

Supplier: Ambeed
Warning

A963127-100MG A963127-1G
77106-450EA 13 USD
77106-450 77106-452
2-Chloro-3-methyl-4-(methylsulfonyl)benzoic acid 95%
2-Chloro-3-methyl-4-(methylsulfonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₄S
MDL Number: MFCD28291815
CAS Number: 106904-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR