(S)-2-Amino-3-(4-fluoro-1H-indol-3-yl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-Trp(4-F)-OH

A961942-100MG A961942-250MG
77106-446EA 97.61 USD
77106-446 77106-448
(S)-2-Amino-3-(4-fluoro-1H-indol-3-yl)propanoic acid 95%
(S)-2-Amino-3-(4-fluoro-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂O₂
MW: 222.21 g/mol
MDL Number: MFCD00137730
CAS Number: 106034-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR