(1S,2S)-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine 97%

Supplier: Ambeed
Danger

A960729-1G A960729-250MG
77106-444EA 78.51 USD
77106-444 77614-672
(1S,2S)-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine 97%
(1S,2S)-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂
MW: 170.29 g/mol
MDL Number: MFCD03411209
CAS Number: 53152-68-4
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR