(2-Aminobenzo[d]thiazol-6-yl)methanol 95%

Supplier: Ambeed
Warning

A960431-100MG A960431-1G
77106-440EA 27 USD
77106-440 77106-442
(2-Aminobenzo[d]thiazol-6-yl)methanol 95%
(2-Aminobenzo[d]thiazol-6-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₈N₂OS
MDL Number: MFCD11976758
CAS Number: 106429-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR