3-Chloro-6-(4-chlorophenyl)pyridazine 95%

Supplier: Ambeed
Warning

A954077-1G A954077-5G
77106-400EA 66.84 USD
77106-400 77106-402
3-Chloro-6-(4-chlorophenyl)pyridazine 95%
3-Chloro-6-(4-chlorophenyl)pyridazine

Precaución: for Research Use Only

Formula: C₁₀H₆Cl₂N₂
MDL Number: MFCD06801340
CAS Number: 58059-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR