2-Bromo-4-iodobenzonitrile 96%

Supplier: Ambeed
Warning

A952621-1G A952621-25G A952621-5G
77106-394EA 10.61 USD
77106-394 77106-396 77106-398
2-Bromo-4-iodobenzonitrile 96%
2-Bromo-4-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
MW: 307.914 g/mol
MDL Number: MFCD18391643
CAS Number: 1261776-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR