(1S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

A952034-100MG A952034-1G A952034-250MG
77106-388EA 125 USD
77106-388 77106-390 77106-392
(1S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol ≥98%, ee 99%
(1S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol

Precaución: for Research Use Only

Formula: C₄₈H₃₀O₂
MDL Number: MFCD09027269
CAS Number: 957111-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR