Piperonyloyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Benzodioxole-5-carbonyl chloride

A950332-10G A950332-1G A950332-5G
77106-382EA 303 USD
77106-382 77106-384 77106-386
Piperonyloyl chloride 98%
Piperonyloyl chloride

Precaución: for Research Use Only

Formula: C₈H₅ClO₃
MW: 184.575 g/mol
MDL Number: MFCD00016904
CAS Number: 25054-53-9
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR