3-Amino-2,6-dichlorobenzonitrile 98+%

Supplier: Ambeed
Warning

A950014-100MG A950014-1G A950014-250MG
77106-376EA 6 USD
77106-376 77106-378 77106-380
3-Amino-2,6-dichlorobenzonitrile 98+%
3-Amino-2,6-dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD09264301
CAS Number: 19846-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR