2,9-Dichloro-4,7-diphenyl-1,10-phenanthroline 95%

Supplier: Ambeed
Warning

A948914-100MG A948914-1G A948914-250MG A948914-5G
77106-354EA 26.53 USD
77106-354 77106-356 77106-358 77573-884
2,9-Dichloro-4,7-diphenyl-1,10-phenanthroline 95%
2,9-Dichloro-4,7-diphenyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₂₄H₁₄Cl₂N₂
MW: 401.29 g/mol
MDL Number: MFCD26398601
CAS Number: 1229012-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR