Cyanine 3-NHS ester 98%

Supplier: Ambeed
Warning

A948587-1MG A948587-5MG
77106-350EA 118.83 USD
77106-350 77106-352
Cyanine 3-NHS ester 98%
Cyanine 3-NHS ester

Precaución: for Research Use Only

Formula: C₃₅H₄₁N₃O₁₀S₂
MW: 727.84 g/mol
MDL Number: MFCD29472576
CAS Number: 146368-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR