(1R,2S,5S)-6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A948364-100MG
A948364-5G
77106-346EA
222.79
USD
77106-346
77106-348
(1R,2S,5S)-6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic acid hydrochloride 95%
(1R,2S,5S)-6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₄ClNO₂ MW: 191.65 g/mol |
MDL Number:
MFCD23106119 CAS Number: 1373205-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...