(S)-tert-Butyl 2-hydroxy-3-methylbutanoate 98%

Supplier: Ambeed
Warning

A947583-100MG A947583-250MG A947583-5G A947583-1G
77106-340EA 26.53 USD
77106-340 77106-342 77106-344 77686-977
(S)-tert-Butyl 2-hydroxy-3-methylbutanoate 98%
(S)-tert-Butyl 2-hydroxy-3-methylbutanoate

Precaución: for Research Use Only

Formula: C₉H₁₈O₃
MW: 174.23 g/mol
MDL Number: MFCD04972376
CAS Number: 3519-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR