4-Ethyl-3-fluorophenylboronic acid 95%

Supplier: Ambeed
Warning

A945717-100MG A945717-1G A945717-250MG
77106-334EA 86 USD
77106-334 77106-336 77686-975
4-Ethyl-3-fluorophenylboronic acid 95%
4-Ethyl-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₂
MDL Number: MFCD22491298
CAS Number: 1374302-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR