1,3-Bis(4-hydroxyphenoxy)benzene 95%

Supplier: Ambeed
Warning

Synonyms: Resorcinol Bis(4-hydroxyphenyl) Ether

A940365-10G A940365-1G A940365-100MG A940365-250MG A940365-5G
77106-312EA 356 USD
77106-312 77106-314 77901-022 77901-379 77901-905
1,3-Bis(4-hydroxyphenoxy)benzene 95%
1,3-Bis(4-hydroxyphenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₄
MW: 294.31 g/mol
MDL Number: MFCD00142505
CAS Number: 126716-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR