2-(Dibromomethyl)-4-fluorobenzonitrile 97%

Supplier: Ambeed
Danger

A935599-100MG A935599-1G A935599-250MG A935599-5G
77106-296EA 70 USD
77106-296 77106-298 77106-300 77106-302
2-(Dibromomethyl)-4-fluorobenzonitrile 97%
2-(Dibromomethyl)-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄Br₂FN
MDL Number: MFCD28405050
CAS Number: 1485536-93-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR