Ethyl indane-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl Indane-5-carboxylate

A933551-1G A933551-250MG
77106-292EA 65 USD
77106-292 77106-294
Ethyl indane-5-carboxylate 95%
Ethyl indane-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
MW: 190.238 g/mol
MDL Number: MFCD12179700
CAS Number: 105640-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR