2,4-Dibromo-6-fluoroaniline 98%

Supplier: Ambeed
Warning

A932447-100G A932447-25G A932447-5G
77106-286EA 29 USD
77106-286 77106-288 77106-290
2,4-Dibromo-6-fluoroaniline 98%
2,4-Dibromo-6-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂FN
MDL Number: MFCD00042230
CAS Number: 141474-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR