2-(Benzyloxy)-4-methylbenzaldehyde 95+%
Supplier: Ambeed
|
Warning
|
A924152-10G
A924152-1G
A924152-5G
77106-254EA
186
USD
77106-254
77106-256
77106-258
2-(Benzyloxy)-4-methylbenzaldehyde 95+%
2-(Benzyloxy)-4-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₅H₁₄O₂ |
MDL Number:
MFCD01415825 CAS Number: 154478-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...