3-Chloro-4-(cyclopropylmethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

A920177-100MG A920177-1G A920177-250MG
77106-240EA 35.01 USD
77106-240 77106-242 77106-244
3-Chloro-4-(cyclopropylmethoxy)phenylboronic acid 98%
3-Chloro-4-(cyclopropylmethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BClO₃
MW: 226.46 g/mol
MDL Number: MFCD18383681
CAS Number: 1228181-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR