5-Carboxy-2-ethoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A918541-100MG A918541-1G A918541-250MG
77106-234EA 31.83 USD
77106-234 77106-236 77106-238
5-Carboxy-2-ethoxyphenylboronic acid 97%
5-Carboxy-2-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
MDL Number: MFCD11044923
CAS Number: 1451391-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR