2-(3-Bromo-2-chlorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A916225-1G A916225-250MG
77106-230EA 140 USD
77106-230 77106-232
2-(3-Bromo-2-chlorophenyl)acetic acid 95%
2-(3-Bromo-2-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD18390581
CAS Number: 1261438-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR