8-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine 95+%

Supplier: Ambeed
Warning

A915649-1G A915649-250MG A915649-5G
77106-216EA 59.42 USD
77106-216 77106-218 77106-220
8-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine 95+%
8-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅ClN₄
MW: 168.58 g/mol
MDL Number: MFCD18072716
CAS Number: 1245644-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR