(R)-(1-Methylpiperidin-3-yl)methanol 95%

Supplier: Ambeed
Warning

A915478-100MG A915478-1G A915478-250MG A915478-5G
77106-208EA 20 USD
77106-208 77106-210 77106-212 77106-214
(R)-(1-Methylpiperidin-3-yl)methanol 95%
(R)-(1-Methylpiperidin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MDL Number: MFCD11035735
CAS Number: 205194-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR