4-Nitro-2-(trifluoromethyl)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
A904444-100MG
A904444-1G
A904444-250MG
77106-192EA
14
USD
77106-192
77106-194
77106-196
4-Nitro-2-(trifluoromethyl)phenylboronic acid 98%
4-Nitro-2-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BF₃NO₄ |
MDL Number:
MFCD22375010 CAS Number: 408359-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...