2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine 98%

Supplier: Ambeed
Warning

A903821-100MG A903821-1G A903821-250MG
77106-182EA 14 USD
77106-182 77106-184 77106-186
2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine 98%
2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₇FINO₂
MW: 283.039 g/mol
MDL Number: MFCD22123918
CAS Number: 1034467-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR