3-[(Methylamino)sulfonyl]benzenecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylsulphamoyl)benzoic acid, 3-[(Methylamino)sulphonyl]benzoic acid

A902109-10G A902109-1G A902109-250MG A902109-5G
77106-174EA 242.95 USD
77106-174 77106-176 77106-178 77106-180
3-[(Methylamino)sulfonyl]benzenecarboxylic acid 97%
3-[(Methylamino)sulfonyl]benzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄S
MW: 215.23 g/mol
MDL Number: MFCD02707876
CAS Number: 35623-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR