1-(2,6-Dichloropyridin-4-yl)ethanone 95%

Supplier: Ambeed
Warning

A897981-100MG A897981-250MG
77106-156EA 11 USD
77106-156 77106-158
1-(2,6-Dichloropyridin-4-yl)ethanone 95%
1-(2,6-Dichloropyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
MDL Number: MFCD11847255
CAS Number: 185319-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR