6,8-Dichloroimidazo[1,2-a]pyrazine 95%

Supplier: Ambeed
Warning

A892303-100MG A892303-250MG
77106-142EA 83.82 USD
77106-142 77106-144
6,8-Dichloroimidazo[1,2-a]pyrazine 95%
6,8-Dichloroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.014 g/mol
MDL Number: MFCD18072707
CAS Number: 1245645-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR