[3-(4-Chlorophenoxy)phenyl]methylamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-(4-Chlorophenoxy)phenyl)methanamine hydrochloride, 3-(Aminomethyl)-4-chlorodiphenyl ether hydrochloride, 3-(4-Chlorophenoxy)benzylamine hydrochloride
A889252-100MG
A889252-1G
A889252-250MG
A889252-5G
77106-114EA
37.14
USD
77106-114
77106-116
77106-118
77106-120
[3-(4-Chlorophenoxy)phenyl]methylamine hydrochloride 95%
[3-(4-Chlorophenoxy)phenyl]methylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₂ClNO·ClH MW: 270.154 g/mol |
MDL Number:
MFCD02089409 CAS Number: 1171023-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...