2-[(4-Chloro-1-naphthyloxy)methyl]phenylboronic acid 95%

Supplier: Ambeed
Warning

A884595-1G A884595-250MG A884595-5G
77106-090EA 45 USD
77106-090 77106-092 77106-094
2-[(4-Chloro-1-naphthyloxy)methyl]phenylboronic acid 95%
2-[(4-Chloro-1-naphthyloxy)methyl]phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₄BClO₃
MDL Number: MFCD08705266
CAS Number: 1072951-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR