2-Chloro-5,8-dimethylquinoline-3-carboxaldehyde 97+%

Supplier: Ambeed
Warning

A883122-1G A883122-250MG A883122-5G
77106-084EA 27.99 USD
77106-084 77106-086 77106-088
2-Chloro-5,8-dimethylquinoline-3-carboxaldehyde 97+%
2-Chloro-5,8-dimethylquinoline-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
MDL Number: MFCD02227045
CAS Number: 323196-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR