2-Hydroxyquinolin-4(1H)-one 98%

Supplier: Ambeed
Warning

A882151-10G A882151-25G
77106-074EA 14 USD
77106-074 77106-076
2-Hydroxyquinolin-4(1H)-one 98%
2-Hydroxyquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD18836846
CAS Number: 70254-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR