5,6-Dimethoxyisobenzofuran-1,3-dione 98%

Supplier: Ambeed
Warning

A878979-100MG A878979-1G A878979-250MG
77106-056EA 64.72 USD
77106-056 77106-058 77106-060
5,6-Dimethoxyisobenzofuran-1,3-dione 98%
5,6-Dimethoxyisobenzofuran-1,3-dione

Precaución: for Research Use Only

Formula: C₁₀H₈O₅
MW: 208.16 g/mol
MDL Number: MFCD27924322
CAS Number: 4821-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR