3,5-Dibromo-2-methylthiophene 96%

Supplier: Ambeed
Warning

A877988-1G A877988-25G A877988-5G
77106-050EA 10 USD
77106-050 77106-052 77106-054
3,5-Dibromo-2-methylthiophene 96%
3,5-Dibromo-2-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₄Br₂S
MW: 255.9583 g/mol
MDL Number: MFCD07781197
CAS Number: 29421-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR