1-(2-Bromo-4-methylthiazol-5-yl)ethanone 97%

Supplier: Ambeed
Warning

A877150-100MG A877150-1G A877150-250MG
77106-044EA 30.77 USD
77106-044 77106-046 77106-048
1-(2-Bromo-4-methylthiazol-5-yl)ethanone 97%
1-(2-Bromo-4-methylthiazol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆BrNOS
MW: 220.08 g/mol
MDL Number: MFCD19370713
CAS Number: 1093106-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR