4-Bromo-3-chlorobenzyl Alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-3-Chlorophenyl)methanol, 4-Bromo-3-chlorobenzyl alcohol

A875629-1G A875629-250MG A875629-5G
77106-038EA 18.04 USD
77106-038 77106-040 77106-042
4-Bromo-3-chlorobenzyl Alcohol 95%
4-Bromo-3-chlorobenzyl Alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.479 g/mol
MDL Number: MFCD08059111
CAS Number: 120077-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR