2-Bromo-4,6-dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A875284-100MG A875284-250MG
77106-034EA 21 USD
77106-034 77106-036
2-Bromo-4,6-dimethoxybenzaldehyde 98%
2-Bromo-4,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD11110636
CAS Number: 81574-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR