5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dichloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine

A872104-100MG A872104-1G A872104-250MG
77106-026EA 40.32 USD
77106-026 77106-028 77106-030
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine 97%
5,7-Dichloro[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₄
MW: 203.029 g/mol
MDL Number: MFCD18642394
CAS Number: 1124383-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR