2-Bromo-6-fluoro-4-nitroaniline 97%

Supplier: Ambeed
Warning

A872022-10G A872022-1G A872022-5G
77106-020EA 90 USD
77106-020 77106-022 77106-024
2-Bromo-6-fluoro-4-nitroaniline 97%
2-Bromo-6-fluoro-4-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFN₂O₂
MDL Number: MFCD09800780
CAS Number: 455-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR