4-Fluoroindoline 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-4-fluoro-1H-indole

A868082-250MG A868082-5G
77105-992EA 71.09 USD
77105-992 77105-994
4-Fluoroindoline 95%
4-Fluoroindoline

Precaución: for Research Use Only

Formula: C₈H₈FN
MW: 137.154 g/mol
MDL Number: MFCD07371634
CAS Number: 552866-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR