α-Chloro-3-fluorotoulene 95%

Supplier: Ambeed
Danger

Synonyms: 1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride

A867662-10G A867662-25G
77105-986EA 7 USD
77105-986 77105-988 77105-990
α-Chloro-3-fluorotoulene 95%
α-Chloro-3-fluorotoulene

Precaución: for Research Use Only

Formula: C₇H₆ClF
MDL Number: MFCD00000904
CAS Number: 456-42-8
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR