Tetrakis(4-hydroxyphenyl)ethylene 98%

Supplier: Ambeed
Warning

A864899-1G A864899-250MG A864899-5G
77105-982EA 111.4 USD
77105-982 77105-984 77614-554
Tetrakis(4-hydroxyphenyl)ethylene 98%
Tetrakis(4-hydroxyphenyl)ethylene

Precaución: for Research Use Only

Formula: C₂₆H₂₀O₄
MW: 396.44 g/mol
MDL Number: MFCD28901376
CAS Number: 119301-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR