3-Chloro-6-(1-ethoxyvinyl)pyridazine 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A863267-1G A863267-250MG
77105-978EA 138.98 USD
77105-978 77105-980
3-Chloro-6-(1-ethoxyvinyl)pyridazine 95% stabilized
3-Chloro-6-(1-ethoxyvinyl)pyridazine

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O
MW: 184.62 g/mol
MDL Number: MFCD27924311
CAS Number: 479628-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR